Assay Details
| Gene Symbol: AT3G23200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113221.3 | NP_188961.2 | 1 - 2 | 451 | 85 | |
| GenBank mRNA | AY128308.1 | - | 1 - 2 | 426 | 85 | |
| BT006540.1 | - | 1 - 2 | 130 | 85 | ||
More Information
Set Membership: |
|

AT3G23200
Genomic Map