Assay Details
| Gene Symbol: PHO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113246.5 | NP_188985.2 | 9 - 10 | 1760 | 94 | |
| GenBank mRNA | AF474076.1 | - | 9 - 10 | 1589 | 94 | |
More Information
Set Membership: |
|

PHO1
Genomic Map