Assay Details
| Gene Symbol: CYN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338643.1 | NP_001327887.1 | 3 - 4 | 686 | 77 | |
| NM_001338644.1 | NP_001327888.1 | 3 - 4 | 873 | 77 | ||
| NM_113252.4 | NP_188991.1 | 3 - 4 | 686 | 77 | ||
| GenBank mRNA | AB004568.1 | - | 3 - 4 | 689 | 77 | |
| AF410298.1 | - | 3 - 4 | 464 | 77 | ||
| AY093708.1 | - | 3 - 4 | 449 | 77 | ||
More Information
Set Membership: |
|

CYN
Genomic Map