Assay Details
Gene Symbol: AT3G23550 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113258.5 | NP_188997.1 | 6 - 7 | 1452 | 77 | |
GenBank mRNA | AY070032.1 | - | 6 - 7 | 1351 | 77 | |
AY096384.1 | - | 6 - 7 | 1302 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
