Assay Details
| Gene Symbol: HGL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125213.2 | NP_001118685.1 | 4 - 5 | 924 | 138 | |
| NM_001338653.1 | NP_001327915.1 | 4 - 5 | 904 | 138 | ||
| NM_113268.3 | NP_566736.1 | 4 - 5 | 647 | 138 | ||
| GenBank mRNA | AY059821.1 | - | 4 - 5 | 532 | 138 | |
| BT002505.1 | - | 4 - 5 | 670 | 138 | ||
| BT008811.1 | - | 3 - 4 | 442 | 138 | ||
More Information
Set Membership: |
|

HGL1
Genomic Map