Assay Details
| Gene Symbol: BAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113297.3 | NP_189034.1 | 2 - 3 | 1346 | 80 | |
| GenBank mRNA | AF367293.1 | - | 2 - 3 | 905 | 80 | |
| AY074393.1 | - | 2 - 3 | 905 | 80 | ||
| AY078046.1 | - | 2 - 3 | 805 | 80 | ||
| AY096517.1 | - | 2 - 3 | 805 | 80 | ||
More Information
Set Membership: |
|

BAM1
Genomic Map