Assay Details
| Gene Symbol: BAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113297.3 | NP_189034.1 | 3 - 4 | 1456 | 77 | |
| GenBank mRNA | AF367293.1 | - | 3 - 4 | 1015 | 77 | |
| AY074393.1 | - | 3 - 4 | 1015 | 77 | ||
| AY078046.1 | - | 3 - 4 | 915 | 77 | ||
| AY096517.1 | - | 3 - 4 | 915 | 77 | ||
More Information
Set Membership: |
|

BAM1
Genomic Map