Assay Details
| Gene Symbol: GR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035679.2 | NP_001030756.2 | 11 - 12 | 1109 | 92 | |
| NM_001125216.1 | NP_001118688.1 | 11 - 12 | 1018 | 92 | ||
| NM_113322.5 | NP_189059.1 | 11 - 12 | 1295 | 92 | ||
| GenBank mRNA | AY140042.1 | - | 11 - 12 | 1076 | 92 | |
| AY142628.1 | - | 10 - 11 | 961 | 92 | ||
| BT008870.1 | - | 10 - 11 | 961 | 92 | ||
| U37697.1 | - | 11 - 12 | 1072 | 92 | ||
More Information
Set Membership: |
|

GR1
Genomic Map