Assay Details
| Gene Symbol: GR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035679.2 | NP_001030756.2 | 13 - 14 | 1283 | 88 | |
| NM_001125216.1 | NP_001118688.1 | 13 - 14 | 1192 | 88 | ||
| NM_113322.5 | NP_189059.1 | 13 - 14 | 1469 | 88 | ||
| GenBank mRNA | AY140042.1 | - | 13 - 14 | 1250 | 88 | |
| AY142628.1 | - | 12 - 13 | 1135 | 88 | ||
| BT008870.1 | - | 12 - 13 | 1135 | 88 | ||
| U37697.1 | - | 13 - 14 | 1246 | 88 | ||
More Information
Set Membership: |
|

GR1
Genomic Map