Assay Details
| Gene Symbol: GR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035679.2 | NP_001030756.2 | 5 - 6 | 500 | 139 | |
| NM_001125216.1 | NP_001118688.1 | 5 - 6 | 409 | 139 | ||
| NM_113322.5 | NP_189059.1 | 5 - 6 | 686 | 139 | ||
| GenBank mRNA | AY140042.1 | - | 5 - 6 | 467 | 139 | |
| AY142628.1 | - | 4 - 5 | 352 | 139 | ||
| BT008870.1 | - | 4 - 5 | 352 | 139 | ||
| U37697.1 | - | 5 - 6 | 463 | 139 | ||
More Information
Set Membership: |
|

GR1
Genomic Map