Assay Details
| Gene Symbol: GR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035679.2 | NP_001030756.2 | 7 - 8 | 782 | 71 | |
| NM_001125216.1 | NP_001118688.1 | 7 - 8 | 691 | 71 | ||
| NM_113322.5 | NP_189059.1 | 7 - 8 | 968 | 71 | ||
| GenBank mRNA | AY140042.1 | - | 7 - 8 | 749 | 71 | |
| AY142628.1 | - | 6 - 7 | 634 | 71 | ||
| BT008870.1 | - | 6 - 7 | 634 | 71 | ||
| U37697.1 | - | 7 - 8 | 745 | 71 | ||
More Information
Set Membership: |
|

GR1
Genomic Map