Assay Details
| Gene Symbol: MSH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338677.1 | NP_001326989.1 | 15 - 16 | 2345 | 74 | |
| NM_113339.4 | NP_189075.2 | 20 - 21 | 2638 | 74 | ||
| GenBank mRNA | AY191303.1 | - | 20 - 21 | 2604 | 74 | |
More Information
Set Membership: |
|

MSH1
Genomic Map