Assay Details
Gene Symbol: AT3G24510 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113361.3 | NP_189094.1 | 1 - 2 | 166 | 72 | |
GenBank mRNA | DQ446690.1 | - | 1 - 2 | 56 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
