Assay Details
| Gene Symbol: ABI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113376.4 | NP_189108.1 | 2 - 3 | 2306 | 68 | |
| GenBank mRNA | X68141.1 | - | 2170 | 68 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ABI3| Gene Symbol: ABI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113376.4 | NP_189108.1 | 2 - 3 | 2306 | 68 | |
| GenBank mRNA | X68141.1 | - | 2170 | 68 | ||
Set Membership: |
|