Assay Details
| Gene Symbol: PRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113392.3 | NP_189124.1 | 1 - 2 | 234 | 118 | |
| GenBank mRNA | AJ224307.1 | - | 1 - 2 | 211 | 118 | |
| AY080799.1 | - | 1 - 2 | 232 | 118 | ||
| AY114077.1 | - | 1 - 2 | 90 | 118 | ||
More Information
Set Membership: |
|

PRT1
Genomic Map