Assay Details
| Gene Symbol: PRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113392.3 | NP_189124.1 | 2 - 3 | 262 | 77 | |
| GenBank mRNA | AJ224307.1 | - | 2 - 3 | 239 | 77 | |
| AY080799.1 | - | 2 - 3 | 260 | 77 | ||
| AY114077.1 | - | 2 - 3 | 118 | 77 | ||
More Information
Set Membership: |
|

PRT1
Genomic Map