Assay Details
| Gene Symbol: ICK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113393.3 | NP_189125.1 | 2 - 3 | 654 | 83 | |
| GenBank mRNA | AJ301556.1 | - | 2 - 3 | 500 | 83 | |
| AK228811.1 | - | 2 - 3 | 594 | 83 | ||
| BT025751.1 | - | 2 - 3 | 500 | 83 | ||
More Information
Set Membership: |
|

ICK3
Genomic Map