Assay Details
| Gene Symbol: FKBP15-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113428.3 | NP_566762.1 | 2 - 3 | 194 | 67 | |
| GenBank mRNA | AY062776.1 | - | 2 - 3 | 147 | 67 | |
| AY128760.1 | - | 2 - 3 | 119 | 67 | ||
More Information
Set Membership: |
|

FKBP15-1
Genomic Map