Assay Details
| Gene Symbol: FKBP15-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113428.3 | NP_566762.1 | 3 - 4 | 252 | 107 | |
| GenBank mRNA | AY062776.1 | - | 3 - 4 | 205 | 107 | |
| AY128760.1 | - | 3 - 4 | 177 | 107 | ||
More Information
Set Membership: |
|

FKBP15-1
Genomic Map