Assay Details
| Gene Symbol: AT3G25280 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113434.1 | NP_189165.1 | 1 - 2 | 97 | 121 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G25280| Gene Symbol: AT3G25280 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113434.1 | NP_189165.1 | 1 - 2 | 97 | 121 | |
Set Membership: |
|