Assay Details
| Gene Symbol: AFH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113446.5 | NP_189177.1 | 2 - 3 | 2338 | 87 | |
| GenBank mRNA | AF174427.1 | - | 2 - 3 | 2113 | 87 | |
| AY080793.1 | - | 2 - 3 | 2183 | 87 | ||
| AY142579.1 | - | 2 - 3 | 2081 | 87 | ||
More Information
Set Membership: |
|

AFH1
Genomic Map