Assay Details
| Gene Symbol: AFH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113446.5 | NP_189177.1 | 3 - 4 | 2584 | 116 | |
| GenBank mRNA | AY080793.1 | - | 3 - 4 | 2429 | 116 | |
| AY142579.1 | - | 3 - 4 | 2327 | 116 | ||
More Information
Set Membership: |
|

AFH1
Genomic Map