Assay Details
| Gene Symbol: ATL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338753.1 | NP_001327862.1 | 4 - 5 | 632 | 84 | |
| NM_113448.5 | NP_566767.1 | 4 - 5 | 512 | 84 | ||
| GenBank mRNA | AY054161.1 | - | 4 - 5 | 421 | 84 | |
| AY065103.1 | - | 4 - 5 | 431 | 84 | ||
| AY081701.1 | - | 3 - 4 | 374 | 84 | ||
| AY136319.1 | - | 4 - 5 | 433 | 84 | ||
| AY186611.1 | - | 3 - 4 | 374 | 84 | ||
| BT000411.1 | - | 3 - 4 | 374 | 84 | ||
| BT002427.1 | - | 4 - 5 | 431 | 84 | ||
| BT008705.1 | - | 3 - 4 | 374 | 84 | ||
More Information
Set Membership: |
|

ATL5
Genomic Map