Assay Details
| Gene Symbol: AOC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113477.5 | NP_566777.1 | 1 - 2 | 451 | 89 | |
| GenBank mRNA | AJ308485.1 | - | 1 - 2 | 321 | 89 | |
| AK229247.1 | - | 1 - 2 | 319 | 89 | ||
| AY087359.1 | - | 1 - 2 | 319 | 89 | ||
More Information
Set Membership: |
|

AOC3
Genomic Map