Assay Details
| Gene Symbol: LTA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113489.3 | NP_189215.1 | 2 - 3 | 1295 | 80 | |
| GenBank mRNA | AF066079.1 | - | 2 - 3 | 1154 | 80 | |
| AY037262.1 | - | 2 - 3 | 1168 | 80 | ||
| BT002343.1 | - | 2 - 3 | 1096 | 80 | ||
More Information
Set Membership: |
|

LTA2
Genomic Map