Assay Details
| Gene Symbol: LTA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113489.3 | NP_189215.1 | 3 - 4 | 1383 | 83 | |
| GenBank mRNA | AF066079.1 | - | 3 - 4 | 1242 | 83 | |
| AY037262.1 | - | 3 - 4 | 1256 | 83 | ||
| BT002343.1 | - | 3 - 4 | 1184 | 83 | ||
More Information
Set Membership: |
|

LTA2
Genomic Map