Assay Details
| Gene Symbol: HMT-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203048.1 | NP_001189977.1 | 1 - 2 | 311 | 79 | |
| NM_113493.5 | NP_189219.1 | 1 - 2 | 390 | 79 | ||
| NM_202632.1 | NP_974361.1 | 1 - 2 | 297 | 79 | ||
| GenBank mRNA | AY065163.1 | - | 1 - 2 | 297 | 79 | |
| AY081604.1 | - | 1 - 2 | 172 | 79 | ||
More Information
Set Membership: |
|

HMT-1
Genomic Map