Assay Details
| Gene Symbol: AT3G25950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113499.2 | NP_189224.1 | 1 | 890 | 55 | |
| GenBank mRNA | AY600580.1 | - | 1 | 890 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G25950| Gene Symbol: AT3G25950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113499.2 | NP_189224.1 | 1 | 890 | 55 | |
| GenBank mRNA | AY600580.1 | - | 1 | 890 | 55 | |
Set Membership: |
|