Assay Details
| Gene Symbol: MAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203049.1 | NP_001189978.1 | 2 - 3 | 389 | 104 | |
| NM_001338771.1 | NP_001327185.1 | 2 - 3 | 502 | 104 | ||
| NM_001338772.1 | NP_001327186.1 | 2 - 3 | 489 | 104 | ||
| NM_113502.3 | NP_189227.1 | 3 - 4 | 401 | 104 | ||
| GenBank mRNA | AK175296.1 | - | 3 - 4 | 401 | 104 | |
| AK176363.1 | - | 3 - 4 | 294 | 104 | ||
| BT010608.1 | - | 3 - 4 | 225 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001338771.1, NM_001338772.1 transcripts.
Set Membership: |
|

MAD2
Genomic Map