Assay Details
| Gene Symbol: MAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203049.1 | NP_001189978.1 | 5 - 6 | 741 | 103 | |
| NM_001338771.1 | NP_001327185.1 | 5 - 6 | 854 | 103 | ||
| NM_001338772.1 | NP_001327186.1 | 5 - 6 | 841 | 103 | ||
| NM_113502.3 | NP_189227.1 | 6 - 7 | 753 | 103 | ||
| GenBank mRNA | AK175296.1 | - | 6 - 7 | 753 | 103 | |
| AK176363.1 | - | 6 - 7 | 646 | 103 | ||
| BT010608.1 | - | 6 - 7 | 577 | 103 | ||
More Information
Set Membership: |
|

MAD2
Genomic Map