Assay Details
| Gene Symbol: AT3G26050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113509.3 | NP_189234.1 | 1 - 2 | 239 | 70 | |
| GenBank mRNA | BT004043.1 | - | 1 - 2 | 194 | 70 | |
| BT004944.1 | - | 1 - 2 | 69 | 70 | ||
More Information
Set Membership: |
|

AT3G26050
Genomic Map