Assay Details
| Gene Symbol: AT3G26050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113509.3 | NP_189234.1 | 3 - 4 | 1180 | 64 | |
| GenBank mRNA | BT004043.1 | - | 3 - 4 | 1135 | 64 | |
| BT004944.1 | - | 3 - 4 | 1010 | 64 | ||
More Information
Set Membership: |
|

AT3G26050
Genomic Map