Assay Details
| Gene Symbol: AT3G26050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113509.3 | NP_189234.1 | 5 - 6 | 1308 | 155 | |
| GenBank mRNA | BT004043.1 | - | 5 - 6 | 1263 | 155 | |
| BT004944.1 | - | 5 - 6 | 1138 | 155 | ||
More Information
Set Membership: |
|

AT3G26050
Genomic Map