Assay Details
| Gene Symbol: EIF4B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113547.2 | NP_189271.1 | 2 - 3 | 1743 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EIF4B1| Gene Symbol: EIF4B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113547.2 | NP_189271.1 | 2 - 3 | 1743 | 83 | |
Set Membership: |
|