Assay Details
| Gene Symbol: TIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113559.4 | NP_189283.1 | 1 - 2 | 588 | 71 | |
| GenBank mRNA | AB493632.1 | - | 1 - 2 | 387 | 71 | |
| AF419613.1 | - | 1 - 2 | 449 | 71 | ||
| AF428341.1 | - | 1 - 2 | 454 | 71 | ||
| AY079114.1 | - | 1 - 2 | 387 | 71 | ||
More Information
Set Membership: |
|

TIP2
Genomic Map