Assay Details
| Gene Symbol: TOM20-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203055.1 | NP_001189984.1 | 3 - 4 | 615 | 102 | |
| NM_113621.3 | NP_189343.1 | 4 - 5 | 499 | 102 | ||
| GenBank mRNA | AJ296023.1 | - | 4 - 5 | 446 | 102 | |
More Information
Set Membership: |
|

TOM20-1
Genomic Map