Assay Details
| Gene Symbol: TOM20-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113622.4 | NP_189344.1 | 1 - 2 | 268 | 124 | |
| GenBank mRNA | AJ296025.1 | - | 1 - 2 | 118 | 124 | |
| AY040027.1 | - | 1 - 2 | 98 | 124 | ||
| AY079411.1 | - | 1 - 2 | 90 | 124 | ||
More Information
Set Membership: |
|

TOM20-3
Genomic Map