Assay Details
| Gene Symbol: CLC-B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113631.4 | NP_189353.1 | 4 - 5 | 1305 | 129 | |
| GenBank mRNA | AY062518.1 | - | 4 - 5 | 1219 | 129 | |
| Z71446.1 | - | 4 - 5 | 1174 | 129 | ||
More Information
Set Membership: |
|

CLC-B
Genomic Map