Assay Details
| Gene Symbol: G6PD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035703.3 | NP_001030780.1 | 15 - 16 | 1604 | 80 | |
| NM_001084745.1 | NP_001078214.1 | 14 - 15 | 1430 | 80 | ||
| NM_001338866.1 | NP_001325679.1 | 14 - 15 | 1567 | 80 | ||
| NM_001338867.1 | NP_001325680.1 | 14 - 15 | 1577 | 80 | ||
| NM_113644.5 | NP_189366.1 | 14 - 15 | 1527 | 80 | ||
| GenBank mRNA | AK317166.1 | - | 15 - 16 | 1499 | 80 | |
| BT021101.1 | - | 13 - 14 | 1336 | 80 | ||
| BT029298.1 | - | 13 - 14 | 1330 | 80 | ||
More Information
Set Membership: |
|

G6PD5
Genomic Map