Assay Details
| Gene Symbol: G6PD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035703.3 | NP_001030780.1 | 4 - 5 | 534 | 141 | |
| NM_001084745.1 | NP_001078214.1 | 3 - 4 | 360 | 141 | ||
| NM_001338866.1 | NP_001325679.1 | 3 - 4 | 497 | 141 | ||
| NM_001338867.1 | NP_001325680.1 | 3 - 4 | 507 | 141 | ||
| NM_113644.5 | NP_189366.1 | 3 - 4 | 457 | 141 | ||
| GenBank mRNA | AK317166.1 | - | 4 - 5 | 429 | 141 | |
| BT021101.1 | - | 2 - 3 | 266 | 141 | ||
| BT029298.1 | - | 2 - 3 | 260 | 141 | ||
More Information
Set Membership: |
|

G6PD5
Genomic Map