Assay Details
| Gene Symbol: G6PD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035703.3 | NP_001030780.1 | 8 - 9 | 1008 | 71 | |
| NM_001084745.1 | NP_001078214.1 | 7 - 8 | 834 | 71 | ||
| NM_001338866.1 | NP_001325679.1 | 7 - 8 | 971 | 71 | ||
| NM_001338867.1 | NP_001325680.1 | 7 - 8 | 981 | 71 | ||
| NM_113644.5 | NP_189366.1 | 7 - 8 | 931 | 71 | ||
| GenBank mRNA | AK317166.1 | - | 8 - 9 | 903 | 71 | |
| BT021101.1 | - | 6 - 7 | 740 | 71 | ||
| BT029298.1 | - | 6 - 7 | 734 | 71 | ||
More Information
Set Membership: |
|

G6PD5
Genomic Map