Assay Details
| Gene Symbol: MOS14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125655.4 | NP_201066.2 | 26 - 27 | 2833 | 151 | |
| GenBank mRNA | AK229829.1 | - | 26 - 27 | 2759 | 151 | |
| AK230415.1 | - | 26 - 27 | 2794 | 151 | ||
| BT002408.1 | - | 26 - 27 | 2783 | 151 | ||
| BT010552.1 | - | 26 - 27 | 2590 | 151 | ||
More Information
Set Membership: |
|

MOS14
Genomic Map