Assay Details
| Gene Symbol: MOS14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125655.4 | NP_201066.2 | 5 - 6 | 701 | 68 | |
| GenBank mRNA | AK229829.1 | - | 5 - 6 | 627 | 68 | |
| AK230415.1 | - | 5 - 6 | 662 | 68 | ||
| BT002408.1 | - | 5 - 6 | 651 | 68 | ||
| BT010552.1 | - | 5 - 6 | 458 | 68 | ||
More Information
Set Membership: |
|

MOS14
Genomic Map