Assay Details
| Gene Symbol: GTR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125663.4 | NP_201074.1 | 1 - 2 | 391 | 114 | |
| GenBank mRNA | AK316918.1 | - | 1 - 2 | 273 | 114 | |
More Information
Set Membership: |
|

GTR2
Genomic Map