Assay Details
| Gene Symbol: GTR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125663.4 | NP_201074.1 | 2 - 3 | 620 | 87 | |
| GenBank mRNA | AK316918.1 | - | 2 - 3 | 502 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GTR2| Gene Symbol: GTR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125663.4 | NP_201074.1 | 2 - 3 | 620 | 87 | |
| GenBank mRNA | AK316918.1 | - | 2 - 3 | 502 | 87 | |
Set Membership: |
|