Assay Details
| Gene Symbol: AT5G62820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125677.4 | NP_201088.1 | 1 - 2 | 716 | 95 | |
| GenBank mRNA | BT022001.1 | - | 1 - 2 | 569 | 95 | |
| BT023479.1 | - | 1 - 2 | 562 | 95 | ||
More Information
Set Membership: |
|

AT5G62820
Genomic Map