Assay Details
| Gene Symbol: AT5G62900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125684.5 | NP_201095.1 | 1 - 2 | 430 | 130 | |
| GenBank mRNA | AF370174.1 | - | 1 - 2 | 324 | 130 | |
| AY133849.1 | - | 1 - 2 | 248 | 130 | ||
More Information
Set Membership: |
|

AT5G62900
Genomic Map