Assay Details
| Gene Symbol: ARR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125686.3 | NP_201097.1 | 2 - 3 | 386 | 85 | |
| GenBank mRNA | AB008489.1 | - | 2 - 3 | 248 | 85 | |
| AK227844.1 | - | 2 - 3 | 280 | 85 | ||
| BT004799.1 | - | 2 - 3 | 226 | 85 | ||
More Information
Set Membership: |
|

ARR6
Genomic Map