Assay Details
| Gene Symbol: BT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345580.1 | NP_001331645.1 | 2 - 3 | 441 | 92 | |
| NM_125711.5 | NP_201121.1 | 2 - 3 | 441 | 92 | ||
| GenBank mRNA | AK227278.1 | - | 2 - 3 | 355 | 92 | |
| AY316674.1 | - | 2 - 3 | 334 | 92 | ||
| BT010872.1 | - | 2 - 3 | 270 | 92 | ||
More Information
Set Membership: |
|

BT1
Genomic Map