Assay Details
| Gene Symbol: DGK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345610.1 | NP_001318872.1 | 1 - 2 | 1870 | 89 | |
| NM_125772.5 | NP_201182.1 | 2 - 3 | 1567 | 89 | ||
| GenBank mRNA | AK228735.1 | - | 2 - 3 | 1330 | 89 | |
| AY062655.1 | - | 2 - 3 | 1322 | 89 | ||
| AY380783.1 | - | 1 - 2 | 1106 | 89 | ||
| BT008792.1 | - | 1 - 2 | 1106 | 89 | ||
More Information
Set Membership: |
|

DGK2
Genomic Map